Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tetrachlorethylen ≥99.5% stabilised

Tetrachlorethylen ≥99.5% stabilised

Glass bottle, 2,5 L
Katalog # 16211-2.5L
Miljø
Irriterende
Sundhed
Leverandør:   Honeywell Chemicals
Tetrachlorethylen ≥99.5% stabilised
Katalog # 16211-2.5L
Leverandør:  Honeywell Chemicals
CAS nummer:  127-18-4
Tetrachlorethylen ≥99.5% stabilised
Tetrachlorethylen ≥99.5% stabilised

Glass bottle, 2,5 L

CAS nummer:  127-18-4
Price
1.240,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents

Beilstein Number: 1361721
RTECS Number: KX3850000
UNSPSC Code: 12190000

Synonymer:  Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
  • CAS nummer:127-18-4
  • Formel:C₂Cl₄
  • MDL nummer:MFCD00000834
  • Molecular Weight:165.83 g/mol
  • Merck Index:12,09332
  • Kogende Pt:120…122 °C (1,013 hPa)
  • Kogende Pt:120-122 °C (1,013 hPa)
  • Smeltende Pt:–22 °C
  • Smeltende Pt:-22 °C
  • Tæthed:1,622…1.624 g/cm3 (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-825-9
  • FN:1897
  • ADR:6.1,III

Documents

Specifikationer

Specifikationstestresultater

  • Vapor Density:
    5.83 (vs air)
  • Contains:
    stabilizer
  • Assay:
    Min. 99.5 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as HCl):
    Max. 0.001 %
  • Refractive Index:
    n20/D 1.505 (lit.)
  • Vapor Pressure:
    19 mmHg ( 25 °C)

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.