Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tetrachlorethylen ≥99.5% stabilised

Tetrachlorethylen ≥99.5% stabilised

Glass bottle, 1 L
Catalog # 16211-1L
Miljø
Irriterende
Sundhed
Supplier:   Honeywell Chemicals
Tetrachlorethylen ≥99.5% stabilised
Catalog # 16211-1L
CAS Number:  127-18-4
Tetrachlorethylen ≥99.5% stabilised
Tetrachlorethylen ≥99.5% stabilised

Glass bottle, 1 L

CAS Number:  127-18-4
Price
718,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

Beilstein Number: 1361721
RTECS Number: KX3850000
UNSPSC Code: 12190000

Synonymer:  Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
  • CAS nummer:127-18-4
  • Formel:C₂Cl₄
  • MDL nummer:MFCD00000834
  • Molecular Weight:165.83 g/mol
  • Merck Index:12,09332
  • Kogende Pt:120…122 °C (1,013 hPa)
  • Kogende Pt:120-122 °C (1,013 hPa)
  • Smeltende Pt:–22 °C
  • Smeltende Pt:-22 °C
  • Tæthed:1,622…1.624 g/cm3 (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-825-9
  • FN:1897
  • ADR:6.1,III

Documents

Specifikationer

Specifikationstestresultater

  • Vapor Density:
    5.83 (vs air)
  • Contains:
    stabilizer
  • Assay:
    Min. 99.5 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as HCl):
    Max. 0.001 %
  • Refractive Index:
    n20/D 1.505 (lit.)
  • Vapor Pressure:
    19 mmHg ( 25 °C)

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer