Buffer solutions, according to DIN 19266, Fluka™
4,008, —, Phthalat, 25 °C, Plastic bottle, 1×1 L, 1 L
Leverandør: Honeywell Chemicals
Buffer solutions, according to DIN 19266, Fluka™
Katalog # 33593-1L
Leverandør: Honeywell Chemicals
CAS nummer:
Gå til:
Product Details & Documents
Buffers with fungicide (except pH 10,012)
: According to DIN 19266.
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Plastic bottle
- pH buffer type:Phthalat
- Pakn:1 L
- Referencetemperatur:25 °C
- pH nøjagtighed:—
- Pk Info:1×1 L
- pH-værdi:4,008



