Behensyre
Glass bottle for solids, 500 mg
Katalog # CAYM9000338-500
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: Docosanoic acid
- CAS nummer:112-85-6
- Formel:C₂₂H₄₄O₂
- MDL nummer:MFCD00002807
- Molecular Weight:340.59 g/mol
- Merck Index:13,01024
- Kogende Pt:306 °C (1013 hPa)
- Smeltende Pt:80 °C
- Tæthed:0,8221 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-010-8
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 mg