Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(R)-(-)-Mandelsyre

(R)-(-)-Mandelsyre

Glass bottle for solids, 50 g
Katalog # CAYM10007128-50
Ætsende
Leverandør:   Cayman Chemical
(R)-(-)-Mandelsyre
Katalog # CAYM10007128-50
Leverandør:  Cayman Chemical
CAS nummer:  611-71-2
(R)-(-)-Mandelsyre

Glass bottle for solids, 50 g

CAS nummer:  611-71-2
Price
1.370,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents
Synonymer:  D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
  • CAS nummer:611-71-2
  • Formel:C₈H₈O₃
  • MDL nummer:MFCD00064251
  • Molecular Weight:152.15 g/mol
  • Kogende Pt:322 °C (1013 hPa)
  • Smeltende Pt:130…132 °C
  • Tæthed:1,341 g/cm³ (20 °C)
  • Flash Pt:>190 °C
  • EINECS:210-276-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.