(R)-(-)-Mandelsyre
Glass bottle for solids, 25 g
Katalog # CAYM10007128-25
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
- CAS nummer:611-71-2
- Formel:C₈H₈O₃
- MDL nummer:MFCD00064251
- Molecular Weight:152.15 g/mol
- Kogende Pt:322 °C (1013 hPa)
- Smeltende Pt:130…132 °C
- Tæthed:1,341 g/cm³ (20 °C)
- Flash Pt:>190 °C
- EINECS:210-276-6
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



