8-Bromisoquinolin
Glass bottle for solids, 1 g
Katalog # CAYM10007082-1
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
- CAS nummer:63927-22-0
- Formel:C₉H₆BrN
- MDL nummer:MFCD04973298
- Molecular Weight:208.0600 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g



