Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
6-Bromisoquinolin

6-Bromisoquinolin

Glass bottle for solids, 1 g
Katalog # CAYM10007080-1
Leverandør:   Cayman Chemical
6-Bromisoquinolin
Katalog # CAYM10007080-1
Leverandør:  Cayman Chemical
CAS nummer:  34784-05-9
6-Bromisoquinolin

Glass bottle for solids, 1 g

CAS nummer:  34784-05-9
Price
704,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
  • CAS nummer:34784-05-9
  • Formel:C₉H₆BrN
  • MDL nummer:MFCD04973299
  • Molecular Weight:208.06 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.