Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Latrunculin A

Latrunculin A

25 µG
Katalog # CAYM10010630-25
Leverandør:   Cayman Chemical
Latrunculin A
Katalog # CAYM10010630-25
Leverandør:  Cayman Chemical
CAS nummer:  76343-93-6
Latrunculin A

25 µG

CAS nummer:  76343-93-6
Price
670,00 kr
Log ind for tilgængelighed
1 * 25 µl
Product Details & Documents

Latrunculin A is a bioactive 2-thiazolidinone macrolide derived from sponges that sequesters G-actin and prevents F-actin assembly. It binds monomeric actin with 1:1 stoichiometry and can be used to block actin polymerization both in vitro (Kd = 0,2 μM) and in cells (0,5 μM, 30 min).

Latrunculin A (1-10 μM) causes depolymerization of tumor cell cytoskeleton within ten minutes. Overnight treatment of cells with latrunculin A (10 μM) strongly suppresses actin synthesis. Prolonged cell treatment blocks dexamethasone-induced changes in actin cytoskeleton with no effect on cell viability. Actin disruption is used to study cell functions in vitro (e.g., migration, endocytosis) and in vivo (e.g., tumor cell invasion).

Synonymer:  (4R)-4-[(1R,4S,5E,7Z,11E,15R,17R)-17-hydroxy-4,11-dimethyl-13-oxo-14,18-dioxabicyclo[13.3.1]nonadeca-5,7,11-trien-17-yl]-1,3-thiazolidin-2-one
  • CAS nummer:76343-93-6
  • Formel:C₂₂H₃₁NO₅S
  • Molecular Weight:421.552 g/mol
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakn:
    25 µG

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.