Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dye terminator removal, Mag-Bind® SeqDTR™

Dye terminator removal, Mag-Bind® SeqDTR™

Mag-Bind™ SeqDTR™, 50 mL
Katalog # M1300-08
Leverandør:   OMEGA BIO-TEK
Dye terminator removal, Mag-Bind® SeqDTR™
Katalog # M1300-08
Leverandør:  OMEGA BIO-TEK
CAS nummer:  
Dye terminator removal, Mag-Bind® SeqDTR™
Dye terminator removal, Mag-Bind® SeqDTR™

Mag-Bind™ SeqDTR™, 50 mL

CAS nummer:  
Price
12.000,00 kr
Log ind for tilgængelighed
1 * 50 mL
Product Details & Documents

Mag-Bind® SeqDTR™ is designed for the effective and reliable removal of unincorporated terminators from sequencing reaction. Sequencing products are first mixed with the Mag-Bind® SeqDTR™. DNA then selectively binds to the Mag-Bind® SeqDTR™ particles. With two rapid wash steps, trace contaminants such as nucleotides, primers and small, non targeted amplification products are removed.

Mag-Bind® SeqDTR™ can be processed in 96- and 384-well formats and is compatible with many liquid handling instruments.

Specifikationer

Specifikationer

  • Pakn:
    50 mL
  • Beskrivelse:
    Mag-Bind™ SeqDTR™
  • Cat. No.:
    M1300-08
  • Prep size:
    Scalable
  • Beskrivelse:
    Mag-Bind™ SeqDTR™
  • Refine target molecule:
    Amplified/modified DNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads
  • Sample type:
    PCR and other enzymatic reactions
  • Sample size:
    Variable

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.