VWR®, Lynlåsposer
80×60 mm
Catalog # 129-0311
Supplier: VWR Collection
Jump to:
Product Details & Documents
Zip bags in LDPE, for storage and transportation of solid samples, tubes, spare parts and documents, available with and without write-on panel.
- Nem og enkel forsegling
- Transparent PE gør det let at identificere indholdet
- Kan lukkes igen flere gange, er fleksibel og kan ikke rives i stykker
Packaging: 100/bag, 10 bags/case.
Documents
Specifications
- L×B:80×60 mm
- Seq:22