Orders LinkContactUsLinkComponent
VWR®, Lynlåsposer
VWR®, Lynlåsposer
60×40 mm
Katalog # 129-0293
Leverandør:   VWR Collection
VWR®, Lynlåsposer
Katalog # 129-0293
Leverandør:  VWR Collection
VWR®, Lynlåsposer
VWR®, Lynlåsposer
60×40 mm
Price
375,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Zip bags in LDPE, for storage and transportation of solid samples, tubes, spare parts and documents, available with and without write-on panel.
  • Nem og enkel forsegling
  • Transparent PE gør det let at identificere indholdet
  • Kan lukkes igen flere gange, er fleksibel og kan ikke rives i stykker
Emballering: 100/bag, 10 bags/case.
Specifikationer
  • L×B:
    60×40 mm
  • Seq:
    01