Orders LinkContactUsLinkComponent
VWR®, Lynlåsposer
VWR®, Lynlåsposer
250×180 mm
  129-0323
VWR®, Lynlåsposer
  129-0323
VWR®, Lynlåsposer
VWR®, Lynlåsposer
250×180 mm
3.040,00 kr
1 * 1.000 Items
Zip bags in LDPE, for storage and transportation of solid samples, tubes, spare parts and documents, available with and without write-on panel.
  • Nem og enkel forsegling
  • Transparent PE gør det let at identificere indholdet
  • Kan lukkes igen flere gange, er fleksibel og kan ikke rives i stykker
 : 100/bag, 10 bags/case.
  • L×B:
    250×180 mm
  • Seq:
    53