Orders LinkContactUsLinkComponent
VWR®, Lynlåsposer
VWR®, Lynlåsposer
170×120 mm
Catalog # 129-0321
Supplier:   VWR Collection
VWR®, Lynlåsposer
Catalog # 129-0321
Supplier:  VWR Collection
VWR®, Lynlåsposer
VWR®, Lynlåsposer
170×120 mm
Price
2.320,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents
Zip bags in LDPE, for storage and transportation of solid samples, tubes, spare parts and documents, available with and without write-on panel.
  • Nem og enkel forsegling
  • Transparent PE gør det let at identificere indholdet
  • Kan lukkes igen flere gange, er fleksibel og kan ikke rives i stykker
Packaging: 100/bag, 10 bags/case.
Specifications
  • L×B:
    170×120 mm
  • Seq:
    51