Orders LinkContactUsLinkComponent
Single channel pipettes, variable volume
2 - 20 µl
Katalog # 613-1357
Leverandør:   EDVOTEK
Single channel pipettes, variable volume
Katalog # 613-1357
Leverandør:  EDVOTEK
Single channel pipettes, variable volume
Single channel pipettes, variable volume
2 - 20 µl
Price
853,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Sturdily designed handheld pipettes with volumes ranging from 0,1 to 5000 µl. They are easy to use, accurate and use standard micropipette tips.
  • Volume easily selected by twisting the top
  • Lightweight design and tip ejector makes operation fast and easy
  • Tool and instructions included for self-calibration
Specifikationer
  • Volumen:
    2 - 20 µl