Orders LinkContactUsLinkComponent
Digested lambda DNA
Lambda DNA digested with Eco RI and Hind III, 20 µG
Katalog # EDVO710
Leverandør:   EDVOTEK
Digested lambda DNA
Katalog # EDVO710
Leverandør:  EDVOTEK
CAS nummer:  
Digested lambda DNA
Lambda DNA digested with Eco RI and Hind III, 20 µG
CAS nummer:  
Price
404,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
20 µg for 20 gels.
Specifikationer
Specifikationer
  • Pakn:
    20 µG
  • Beskrivelse:
    Lambda DNA digested with Eco RI and Hind III