Orders LinkContactUsLinkComponent
Spin Adapters for Gravity Columns
Spin Adapters for Gravity Columns
MidiSpin™ adapter, for PD MidiTrap™ G-25 columns
Katalog # 28-9232-44
Leverandør:   Cytiva
Spin Adapters for Gravity Columns
Katalog # 28-9232-44
Leverandør:  Cytiva
Spin Adapters for Gravity Columns
Spin Adapters for Gravity Columns
MidiSpin™ adapter, for PD MidiTrap™ G-25 columns
Price
150,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
  • Convenient adapters, enable spinning of all gravity columns
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
Specifikationer
  • Beskrivelse:
    MidiSpin™ adapter, for PD MidiTrap™ G-25 columns
  • Seq:
    04