Affinity chromatography columns, HiTrap™ MabSelect SuRe™
HiTrap™ MabSelect SuRe, PP, 85 µm, 7×25 mm, 1 ml, 4 ml/min
Leverandør: Cytiva
Affinity chromatography columns, HiTrap™ MabSelect SuRe™
Katalog # 11-0034-93
Leverandør: Cytiva
Gå til:
Product Details & Documents
HiTrap MabSelect SuRe columns are prepacked with MabSelect SuRe, a BioProcess resin for purification of monoclonal antibodies from large sample volumes.
- Withstands rigorous CIP and sanitization procedures with 0,1 to 0,5 M NaOH
- Novel ligand design gives enhanced protease resistance resulting in lower ligand leakage
- Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
- Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs
The novel alkali-tolerant recombinant Protein A ligand allow cleaning-in-place (CIP).
Documents
Certifikater
Søg efter partinummer11-0034-93_42 br 11-0034-93_42 br (1) 11-0034-93_42 DF 11-0034-93_42 DF (1) 11-0034-93_42 HB
Datasheet
More
11-0034-93_42 IN 11003493_HAND BOOK 11003493_HAND BOOK 1 11003493_INSTRUCTIONS 11003493_INSTRUCTIONS 1
Manual
More
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifikationer
- Flowhastighed:4 ml/min
- Partikelstørrelse:85 µm
- Kapacitet:1 ml
- Beskrivelse:HiTrap™ MabSelect SuRe
- Seq:02
- Kolonnemateriale:PP
- Sengens dimensioner:7×25 mm
- Cat. No.:11-0034-93
- Opbevaring:2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
- Maks. tryk:5 bar
- pH-område:3 - 12
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



