Affinity chromatography columns, HiTrap™ MabSelect SuRe™
HiTrap™ MabSelect SuRe, PP, 85 µm, 16×25 mm, 5 ml, 20 ml/min
Katalog # 11-0034-95
Leverandør: Cytiva
Affinity chromatography columns, HiTrap™ MabSelect SuRe™
Katalog # 11-0034-95
Leverandør: Cytiva
Gå til:
Product Details & Documents
HiTrap MabSelect SuRe columns are prepacked with MabSelect SuRe, a BioProcess resin for purification of monoclonal antibodies from large sample volumes.
- Withstands rigorous CIP and sanitization procedures with 0,1 to 0,5 M NaOH
- Novel ligand design gives enhanced protease resistance resulting in lower ligand leakage
- Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
- Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs
The novel alkali-tolerant recombinant Protein A ligand allow cleaning-in-place (CIP).
Documents
Certifikater
Søg efter partinummer11-0034-93_42 br 11-0034-93_42 br (1) 11-0034-93_42 DF 11-0034-93_42 DF (1) 11-0034-93_42 HB
Datablad
More
11-0034-93_42 IN 11003493_HAND BOOK 11003493_HAND BOOK 1 11003493_INSTRUCTIONS 11003493_INSTRUCTIONS 1
Manuel
More
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Yderligere produktdetaljer
Specifikationer
- Flowhastighed:20 ml/min
- Partikelstørrelse:85 µm
- Kapacitet:5 ml
- Beskrivelse:HiTrap™ MabSelect SuRe
- Seq:04
- Kolonnemateriale:PP
- Sengens dimensioner:16×25 mm
- Cat. No.:11-0034-95
- Opbevaring:2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)
- Maks. tryk:5 bar
- pH-område:3 - 12



