Orders LinkContactUsLinkComponent
dNTP solutions, illustra™
dNTP solutions, illustra™
dUTP, 100 mM solution, 25 µmol
Catalog # 28-4065-41
Supplier:   Cytiva
dNTP solutions, illustra™
Catalog # 28-4065-41
Supplier:  Cytiva
CAS Number:  
dNTP solutions, illustra™
dNTP solutions, illustra™
dUTP, 100 mM solution, 25 µmol
CAS Number:  
Price
398,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Specifications
Specifications
  • Pakn:
    25 µmol
  • Beskrivelse:
    dUTP, 100 mM solution
Specifications