Accessories
Selector disc, black, G 7855
Catalog # 141-0842
Supplier: MIELE
Jump to:
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifications
- Beskrivelse:Selector disc, black
- Seq:32
- Til:G 7855