Wipers for general applications, PIG®
Servietter, allround, i pop-up-æske, hvid, 420×240 mm, 75 servietter/æske
Katalog # NEWPWIP311
Leverandør: NEW PIG
Gå til:
Product Details & Documents
These cellulose wipes with adhesive latex binder are ideal for all kinds of jobs - from cleaning parts to cleaning hands.
- Constructed of soft, absorbent cellulose fibers with a high-quality binder to increase strength for wet or dry wiping
- Made thick and absorbent to quickly soak up oils, degreasers and other liquids
- Low-lint material won't leave residue or create a mess
- Excellent for a variety of wiping applications from quick cleanups to degreasing
- Available 1/4 folded and poly wrapped, as economical rolls or in convenient pop-up boxes
Specifikationer
- Pakket:75 servietter/æske
- Farve:hvid
- Beskrivelse:Servietter, allround, i pop-up-æske
- L×B:420×240 mm
- Seq:06