Orders LinkContactUsLinkComponent
Wipers for general applications, PIG®
Wipers for general applications, PIG®
Servietter, allround, 1/4 foldede, blå, 330×300 mm, 56 servietter/pakke
Katalog # NEWPWIP230
Leverandør:   NEW PIG
Wipers for general applications, PIG®
Katalog # NEWPWIP230
Leverandør:  NEW PIG
Wipers for general applications, PIG®
Wipers for general applications, PIG®
Servietter, allround, 1/4 foldede, blå, 330×300 mm, 56 servietter/pakke
Price
1.190,00 kr
Log ind for tilgængelighed
1 * 56 Items
Product Details & Documents
These cellulose wipes with adhesive latex binder are ideal for all kinds of jobs - from cleaning parts to cleaning hands.
  • Constructed of soft, absorbent cellulose fibers with a high-quality binder to increase strength for wet or dry wiping
  • Made thick and absorbent to quickly soak up oils, degreasers and other liquids
  • Low-lint material won't leave residue or create a mess
  • Excellent for a variety of wiping applications from quick cleanups to degreasing
  • Available 1/4 folded and poly wrapped, as economical rolls or in convenient pop-up boxes
Specifikationer
  • Pakket:
    56 servietter/pakke
  • Farve:
    blå
  • Beskrivelse:
    Servietter, allround, 1/4 foldede
  • L×B:
    330×300 mm
  • Seq:
    01