PIG® Overpacks
Overpack, UN1H2/X70/S, 115 L, PP, 425×690 (inner) mm, hvid, 61 L drum, 6,8 kg
Katalog # NEWPPAK709-WH
Leverandør: NEW PIG
Gå til:
Product Details & Documents
Tough enough for rough handling, guaranteed. Ship your haz waste in our UN rated overpacks to stay compliant with regs.
- Containers are nestable when empty to save storage space
- Designed and proven to withstand the abuse of shipping to help keep hazardous waste safely contained
- Material with UV inhibitors is lightweight and chemical-resistant
X-rated for packing groups I, II and III, so it's ideal for regulated packaging applications, including shipping hazardous waste by land, sea or rail.
Advarsel: If using this product with flammable liquids, please consider the regulations that apply to storage and handling of flammable liquids and the safety of this application, specifically flammable vapours, static discharge and heat sources.
Specifikationer
- Diameter×H:425×690 (inner) mm
- Kapacitet:115 L
- Farve:hvid
- Beskrivelse:Overpack, UN1H2/X70/S
- Materiale:PP
- Seq:02
- Vægt:6,8 kg
- Max. fodermængden:61 L drum



