PIG® poly spill containment pallets
4-drum poly spill containment pallet, 250 L, 1295×1295×254 mm
Catalog # 121-0188
Supplier: NEW PIG
Gå til:
Product Details & Documents
These economical pallets are strong enough to handle long-term storage of fully loaded steel drums.
- Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
- Molded-in sump catches leaks, drips and spills to help you comply with regulations and keep your storage areas clean and safe
- Strong enough to support four fully loaded steel or poly drums
- Textured grating adds traction and lifts out for easy sump access
- 2-way forklift entry allows easy movement from either direction (unloaded only)
Nestable design permits stacking when not in use; lowers cost of bulk shipping.
Specifikationer
- Kapacitet:250 L
- Beskrivelse:4-drum poly spill containment pallet
- B×D×H:1295×1295×254 mm



