Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
PIG® spill bag

PIG® spill bag

Spill bag, includes 20 mat pads, 1 absorbent sock and 2 disposal bags, 11,3 L, 140×250×360 mm
Katalog # 121-0074
Leverandør:   NEW PIG
PIG® spill bag
Katalog # 121-0074
Leverandør:  NEW PIG
PIG® spill bag
PIG® spill bag

Spill bag, includes 20 mat pads, 1 absorbent sock and 2 disposal bags, 11,3 L, 140×250×360 mm

Price
363,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

High-visibility, compact spill bag travels anywhere you do for spill response on the go.

  • Brightly coloured yellow bag features a reflective stripe for greater visibility and easy identification
  • Drawstring closure is ideal for carrying or hanging on a wall
  • Packed with a variety of PIG® absorbents to handle small spills

PIG® spill bag is flexible and small enough to fit behind the seat of a lorry; stows almost anywhere and is easy to carry in the event of a spill

Advarsel: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.

Specifikationer
  • Beskrivelse:
    Spill bag, includes 20 mat pads, 1 absorbent sock and 2 disposal bags
  • Seq:
    003
  • B×D×H:
    140×250×360 mm
  • Sugeevne:
    11,3 L