Orders LinkContactUsLinkComponent
PIG® medium and large mobile container kits
Spill kit, Oil-Only, includes 120 mat pads, 8 socks, 12 pillows and 10 disposal bags, 168,3 L, 730×570×1050 mm
Katalog # 121-0072
Leverandør:   NEW PIG
PIG® medium and large mobile container kits
Katalog # 121-0072
Leverandør:  NEW PIG
PIG® medium and large mobile container kits
PIG® medium and large mobile container kits
Spill kit, Oil-Only, includes 120 mat pads, 8 socks, 12 pillows and 10 disposal bags, 168,3 L, 730×570×1050 mm
Price
4.550,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Fast, effective response to indoor and outdoor spills.
  • Durable, wheeled 240 and 360 L container is packed with high-quality PIG® absorbents
  • Easy to transport supplies directly to the spill site
  • Hinged lid makes it easy to get the absorbents you need — just flip it open
  • Container with bold Spill Kit lettering for quick identification
  • Absorbents are organised for quick, easy response
  • Heavy-duty rubber wheels roll easily over nearly any surface
Weather-resistant polyethylene container protects the contents from moisture, dirt and damage (containers are best kept indoors for long-term storage).
Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Specifikationer
  • Beskrivelse:
    Spill kit, Oil-Only, includes 120 mat pads, 8 socks, 12 pillows and 10 disposal bags
  • Seq:
    05
  • B×D×H:
    730×570×1050 mm
  • Sugeevne:
    168,3 L