Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
PIG® BLUE absorbent socks

PIG® BLUE absorbent socks

Absorbent socks, PP skin/vermiculate filler, 56 L, 20 strømper/pose, 80×1220 mm
Katalog # NEWP2048
Leverandør:   NEW PIG
PIG® BLUE absorbent socks
Katalog # NEWP2048
Leverandør:  NEW PIG
PIG® BLUE absorbent socks
PIG® BLUE absorbent socks

Absorbent socks, PP skin/vermiculate filler, 56 L, 20 strømper/pose, 80×1220 mm

Price
883,00 kr
Log ind for tilgængelighed
1 * 20 Items
Product Details & Documents

Surround spills to prevent spreading. Extra-fine vermiculite filler pulls in liquid and hugs floors to stay in place.

  • Heaviest sock, features extra-dense construction to hug floors and stay in place whilst containing spills
  • Fine-grade vermiculite filler quickly absorbs leaks, drips and spills; great for spill response or absorbing machine leaks
  • Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water

Ideal for diking and damming liquids. Can be air-dried for reuse when soaked with water.

Specifikationer
  • Beskrivelse:
    Absorbent socks, PP skin/vermiculate filler
  • Seq:
    03
  • Sugeevne:
    56 L
  • Pakket:
    20 strømper/pose
  • Ø×L:
    80×1220 mm