Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, TEMPase Hot Start 2× master mix

Blue TEMPase Hot Start 2X Master Mix, med Blue Master Mix C, 1,5 mM MgCl₂, 2.500 Tests
Katalog # 733-1841
Leverandør:   VWR Chemicals
VWR®, TEMPase Hot Start 2× master mix
Katalog # 733-1841
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, TEMPase Hot Start 2× master mix
VWR®, TEMPase Hot Start 2× master mix

Blue TEMPase Hot Start 2X Master Mix, med Blue Master Mix C, 1,5 mM MgCl₂, 2.500 Tests

CAS nummer:  
Price
19.100,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

TEMPase Hot Start DNA Polymerase Master Mix and Blue TEMPase Master Mix are good alternatives to TEMPase Hot Start DNA Polymerase. These master mixes offer easy reaction assembly at room temperature, reduced set-up time and fewer handling steps, which lead to increased reproducibility. As a consequence TEMPase Hot Start DNA Polymerase Master Mix is highly suited to standard tests.

The blue loading dye in Blue TEMPase Hot Start DNA Polymerase Master Mix facilitates direct gel loading, and eliminates the need for separate loading dye - no need for time-consuming sample preparation before electrophoresis.

Multiplex 2× Master Mix is composed of TEMPase Hot Start DNA Polymerase and a specialised buffer system designed for multiplex PCR.

 : TEMPase Hot Start DNA Polymerase Master Mix and Blue TEMPase Master Mix are available in two variations, either based on Key Buffer (Master Mix K) or Combination Buffer (Master Mix C) to suit different PCR requirements. Additional MgCl₂ is included in the kit to enable optimisation.

Specifikationer

Specifikationer

  • Pakn:
    2.500 Tests
  • Beskrivelse:
    Blue TEMPase Hot Start 2X Master Mix, med Blue Master Mix C, 1,5 mM MgCl₂
  • Høj troskab:
    No
  • Enheder:
    2500 r Mix 2500
  • Funktioner:
    Direct loading
  • Exonuklease aktivitet:
    5’→3’
  • Koncentration:
    2x
  • Buffer sammensætning:
    Based on Combination buffer
  • Størrelse:
    50 x 1.25 ml
  • Cat. No.:
    733-1841
  • Beskrivelse:
    Blue TEMPase Hot Start DNA Polymerase 2X Master Mix C<br />1.5 mM MgCl2 2500 reactions
  • Format:
    Mastermix incl. dNTPs
  • Ansøgning:
    Multiplex
  • Amplikon størrelse:
    ≤ 5kb
  • Processivity:
    Standard
  • Product overhang:
    3'A
  • Varmstart:
    Chemically modified
  • Opbevaring betingelser:
    – 20 °C (long term), + 4 °C (6 months)

Specifikationer