Compression fittings
Luer to small hub RN adaptor
Katalog # HAMI55753-01
Leverandør: HAMILTON BONADUZ
Gå til:
Product Details & Documents
The compression fittings and adaptors are designed to connect Hamilton removable needle (RN) and knurled hub (KH) syringes to glass capillary tubing and PEEK tubing. The connectors can be combined in a variety of different configurations to meet most connection needs.
Applications include: Neuroscience injections, patch clamp, cell transplantation, stem cell research, corrosive chemical dispensing, infusion pump injections, and mass spectrometry infusion pumps.
Specifikationer
- Beskrivelse:Luer to small hub RN adaptor
- Seq:04