Orders LinkContactUsLinkComponent
Compression fittings
Compression fittings
Dual small hub RN coupler
Katalog # HAMI55752-01
Leverandør:   HAMILTON BONADUZ
Compression fittings
Katalog # HAMI55752-01
Leverandør:  HAMILTON BONADUZ
Compression fittings
Compression fittings
Dual small hub RN coupler
Price
673,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The compression fittings and adaptors are designed to connect Hamilton removable needle (RN) and knurled hub (KH) syringes to glass capillary tubing and PEEK tubing. The connectors can be combined in a variety of different configurations to meet most connection needs.
Applications include: Neuroscience injections, patch clamp, cell transplantation, stem cell research, corrosive chemical dispensing, infusion pump injections, and mass spectrometry infusion pumps.
Specifikationer
  • Beskrivelse:
    Dual small hub RN coupler
  • Seq:
    03