Plugs for test tubes
LDPE, 16 mm, gul
Katalog # 212-0521
Leverandør: BOETTGER PAUL
Gå til:
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
- Not autoclavable
- Easy handling with a high, ribbed grip
- Easy to open by tilting 'one-hand operation'
Specifikationer
- Farve:gul
- Materiale:LDPE
- Seq:09
- Størrelse:16 mm