Orders LinkContactUsLinkComponent
Test tubes with screw caps
15 ml, 76 mm, 20 mm, Colour-21390331|Colour-21390353, Conical bottom, skirted, screw cap enclosed, non sterile
  212-0511
Test tubes with screw caps
  212-0511
Test tubes with screw caps
Test tubes with screw caps
15 ml, 76 mm, 20 mm, Colour-21390331|Colour-21390353, Conical bottom, skirted, screw cap enclosed, non sterile
1.510,00 kr
1 * 1.000 Items
PP, transparent.
  • Good chemical and temperature resistance
  • Tight sealing screw cap made from PE
  • Available with enclosed or fitted screw cap, standard-hygienic or sterile
For safe transportation and storage of samples.
  • Udv.Ø:
    20 mm
  • Model:
    non sterile
  • Kapacitet:
    15 ml
  • Beskrivelse:
    Conical bottom, skirted, screw cap enclosed
  • Længde:
    76 mm
  • Seq:
    02
  • Lågfarve:
    Colour-21390331|Colour-21390353