Orders LinkContactUsLinkComponent
NMR tubes, polyethylene pressure caps
NMR tubes, polyethylene pressure caps
10 mm, rød
Katalog # SCERSP897095-1001
Leverandør:   DWK Life Sciences
NMR tubes, polyethylene pressure caps
Katalog # SCERSP897095-1001
Leverandør:  DWK Life Sciences
NMR tubes, polyethylene pressure caps
10 mm, rød
Price
352,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.
Specifikationer
  • Farve:
    rød
  • Længde:
    10 mm