NMR tubes, polyethylene pressure caps
5 mm, grøn
Katalog # SCERSP897095-0071
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.
Specifikationer
- Farve:grøn
- Længde:5 mm
- Seq:04