Orders LinkContactUsLinkComponent
NMR tubes, polyethylene pressure caps
NMR tubes, polyethylene pressure caps
5 mm, sort
Katalog # SCERSP897095-0021
Leverandør:   DWK Life Sciences
NMR tubes, polyethylene pressure caps
Katalog # SCERSP897095-0021
Leverandør:  DWK Life Sciences
NMR tubes, polyethylene pressure caps
5 mm, sort
Price
235,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.
Specifikationer
  • Farve:
    sort
  • Længde:
    5 mm
  • Seq:
    02