NMR tubes, polyethylene pressure caps
5 mm, rød
Katalog # SCERSP897095-0001
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.
Specifikationer
- Farve:rød
- Længde:5 mm
- Seq:01