Homogeniser, Dounce type, KIMBLE®
2 ml
Katalog # 432-0262
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Designed primarily for cellular work where the nucleus remains intact after homogenisation.
- All-glass construction
- Two pestles are supplied with each complete unit
- Large clearance pestle is used for the initial sample reduction
- Small clearance pestle is used to form the final homogenate
- Replacement components are available and completely interchangeable
Specifikationer
- Kapacitet:2 ml
- Seq:06