Orders LinkContactUsLinkComponent
Homogeniser, Dounce type, KIMBLE®
Homogeniser, Dounce type, KIMBLE®
100 ml
Katalog # SCERSP885300-0100
Leverandør:   DWK Life Sciences
Homogeniser, Dounce type, KIMBLE®
Katalog # SCERSP885300-0100
Leverandør:  DWK Life Sciences
Homogeniser, Dounce type, KIMBLE®
Homogeniser, Dounce type, KIMBLE®
100 ml
Price
7.200,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Designed primarily for cellular work where the nucleus remains intact after homogenisation.
  • All-glass construction
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable
Specifikationer
  • Kapacitet:
    100 ml
  • Seq:
    05