Micro capillaries, disposable, Kimble®
Capillary pipettes, 50 µl, grøn
Katalog # SCERSP71900-50
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
These disposable micro capillary pipettes are made from soda lime glass, USP Type III.
- Calibrated to contain
- Colour-coded
- Marked with single black capacity ring
- Packaged in easy-open cylinders for convenient dispensing
Certifications: Designed from ASTM Specification E672.
Emballering: Each cylinder contains 250 pipettes and one pipette device.
Specifikationer
- Farvekode:grøn
- Kapacitet:50 µl
- Beskrivelse:Capillary pipettes
- Seq:03