Micro capillaries, disposable, Kimble®
Capillary pipettes, 10 µl, orange
Katalog # SCERSP71900-10
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
These disposable micro capillary pipettes are made from soda lime glass, USP Type III.
- Calibrated to contain
- Colour-coded
- Marked with single black capacity ring
- Packaged in easy-open cylinders for convenient dispensing
Certifications: Designed from ASTM Specification E672.
Emballering: Each cylinder contains 250 pipettes and one pipette device.
Specifikationer
- Farvekode:orange
- Kapacitet:10 µl
- Beskrivelse:Capillary pipettes
- Seq:02