Orders LinkContactUsLinkComponent
High visibility jacket, Strada 2 High-Visibility
High visibility jacket, Strada 2 High-Visibility
gul/blå, L
Katalog # 113-0913
Leverandør:   DELTA PLUS
High visibility jacket, Strada 2 High-Visibility
Katalog # 113-0913
Leverandør:  DELTA PLUS
High visibility jacket, Strada 2 High-Visibility
High visibility jacket, Strada 2 High-Visibility
gul/blå, L
Price
1.090,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This high-visibility safety jacket (class 3) is made from high visibility, fluorescent polyester with internal polyurethane coating and polyester lining. The jacket includes a zip with press stud cover, two pockets with press stud pocket covers, two side pockets, an inside pocket, and waterproof seams. Suitable for ∼50 washes at 40 °C.
  • Two reflective strips on each sleeve and strips on the body and shoulders
  • Non detachable hood with velcro fastening, which can be folded in the collar
  • Straight sleeve openings with wind-resistant, internal elastic cuffs
  • Ventilation eyelets in the underarm area
  • Can be tightened at the waist with elastic cord and toggle
Colour: Fluorescent yellow/marine blue or fluorescent orange/marine blue.
Certifications: EN 340; EN 342; EN 343; EN 471
Specifikationer
  • Tøj størrelse:
    L
  • Farve:
    gul/blå
  • Seq:
    02
  • Køn:
    Unisex
  • Cat. No.:
    113-0913
  • Sleeve or leg length:
    Long
  • Risk area:
    HiVis
  • Outfit applikationer:
    Industrial
  • Lukkestørrelse:
    Med lynlås
  • Stof:
    Polyester with internal polyurethane coating