Inlet liners
Cyclo double gooseneck liner for split operation, 6,2 mm, 4 mm, 92,1
Katalog # PERCN6502005
Leverandør: PerkinElmer
Gå til:
Product Details & Documents
Liners protect GC columns against column contamination. These glass split and splitless inlet liners promote full vaporisation of samples, minimise mass discrimination, and achieve accurate and reproducible results.
- Liners packed with wool help to trap non-volatile material
Specifikationer
- Udv.Ø:6,2 mm
- Ø indv.:4 mm
- Beskrivelse:Cyclo double gooseneck liner for split operation
- Længde:92,1
- Seq:14