Blood collection needles, BD Vacutainer® Eclipse™
21, 32 mm, grøn
Katalog # BDAM368609
Leverandør: BD Medical
Gå til:
Product Details & Documents
The BD Vacutainer® Eclipse™ blood collection needle is a safety-engineered, multi-sample blood collection needle that offers a simple, effective way to collect blood while reducing the possibility of needlestick injuries. It features a safety shield that allows for one-handed activation to cover the needle immediately upon withdrawal from the vein and confirms proper activation with an audible click.
- Built-in safety shield is aligned with the bevel of the needle
- The safety shield permanently locks into place, significantly reducing the risk of needlestick injuries
- The pre-attached holder (only supplied with some models) minimises the risk of accidental needlestick injuries from the holder end of the device
Specifikationer
- Farvekode:grøn
- Måler:21
- Længde:32 mm
- Seq:01