Orders LinkContactUsLinkComponent
PIG® spill caddy
PIG® spill caddy
Refill universal spill caddy, 51,5 L
Katalog # 121-0112
Leverandør:   NEW PIG
PIG® spill caddy
Katalog # 121-0112
Leverandør:  NEW PIG
PIG® spill caddy
PIG® spill caddy
Refill universal spill caddy, 51,5 L
Price
1.160,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
The mobile PIG® spill caddy for fast spill response or daily leak maintenance.
  • High-visibility yellow container is easy to spot when you need it
  • Contents are compartmentalised for easy access
  • Adjustable shelves
  • 20 cm wheels for easy mobility
  • Double-wall polyethylene cart for durability
Container can be locked with a padlock (not included) to prevent pilferage.
Advarsel: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Specifikationer
  • Beskrivelse:
    Refill universal spill caddy
  • Seq:
    02
  • Sugeevne:
    51,5 L