Drum taps, brass
Self-closing with flame guard and PTFE seal, Aggressive liquids, 3/4"
Katalog # MSGA2798
Leverandør: asecos
Gå til:
Product Details & Documents
Brass taps for drums with screw thread.
- Self closing
- Flame guard
- PTFE seal
Bestillingsoplysninger: MSGA2799 includes a trailer fitting for decanting vessels.
Specifikationer
- Gevind:3/4"
- Beskrivelse:Self-closing with flame guard and PTFE seal
- Seq:02
- Til:Aggressive liquids