Universal socks
Universal sorbent snakes, 6 per pack, 75×3000 mm
Katalog # MSGA11347
Leverandør: asecos
Gå til:
Product Details & Documents
Universal socks for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
- Optimal for long term use
- Capacity up to 16 times of its own weight
- High rib and rub resistance
For containment and absorption of larger leaks.
Specifikationer
- Beskrivelse:Universal sorbent snakes
- Pakket:6 per pack
- Ø×L:75×3000 mm