Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Test tube racks, Plexiglas®

33 (3×11), 18 mm, 80×310×80 mm
  HECH42808033
Test tube racks, Plexiglas®
  HECH42808033
Test tube racks, Plexiglas®
Test tube racks, Plexiglas®

33 (3×11), 18 mm, 80×310×80 mm

 
686,00 kr
   
1 * 1 Items
 

Test tube racks, made of Plexiglas®, with numbered holes.

  • Good thermal resistance
  • Rugged stable design
  • Easy to clean
 
  • Til rør Ø:
    18 mm
  • Ant. huller:
    33 (3×11)
  • Seq:
    05
  • B×D×H:
    80×310×80 mm