Handsker, nitril, Kimtech™ Blue Nitrile
S, blå, 240 mm, 100/box
Katalog # 112-4867
Leverandør: KIMBERLY CLARK
Gå til:
Product Details & Documents
These gloves are manufactured from nitrile instead of rubber latex, which reduces the likelihood of TYPE I glove-associated reactions.
- Constructed of synthetic nitrile polymer and therefore free of silicone and latex
- Smooth bisque finish enables excellent tactile sensitivity in the handling of both wet and dry materials and equipment
- Beaded cuffs for added strength and easier donning and doffing
- Compliant protection against viruses, chemical splash and microbiological hazards
- Virus protection according to ISO 16604 procedure B:2004
- Static dissipative in-use and anti-static tested, meeting the requirements defined in EN 1149-5
Suitable for a wide variety of non sterile applications, KIMTECH SCIENCE* Blue Nitrile gloves are designed to protect laboratory processes through minimising contamination risk and boosting productivity. Ideal for use in chemistry, biochemistry, microbiology, virology, life sciences, biomedical research, forensic science and non sterile drug manufacturing.
EN 374, EN 455-1/-2/-3, kat. III, AQL 1,5
Thickness: 0,12 mm (palm), 0,15 mm (finger)
Colour: Blue
EN 374, EN 455-1/-2/-3, Cat. III, AQL 1,5
Certifications: PPE Cat III (EU) 2016/425, EN 374-1 Type C (K), EN 374-5 (VIRUS), AQL 1.5
Documents
BLUE NITRILE*
Billeder 360
Kimberly-Clark Kimtech™ Lab Gloves brochure
Produktlitteratur
Specifikationer
- Handske størrelse:S
- Pakket:100/box
- Farve:blå
- Længde:240 mm
- Seq:01
- Håndflade tykkelse:0,12 mm
- Antistatisk:+
- Trækstyrke:21 MPa
- Med pudder/pudderfri:Pudderfri
- Manchet tykkelse:0,09 mm
- Finger tykkelse:0,15 mm
- Manchet:Rullet kant
- EN norm:EN 374-1 Type C (K), EN 374-5 (VIRUS)
- Cat. No.:112-4867
- Ambidextrous/Hånd-specifik:Ambidextrous
- Latex/latex-fri:Latex-fri
- Engangs/Genbrugelig:Engangs
- Ultimativ forlængelse:550 % (før ældning), 500 % (efter accelereret ældning)



