Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Threaded fittings

Threaded fittings

Tubing nozzle, fix, bend, external thread, nozzle-Ø 16 mm, NW 11 mm, PP, 12,7 mm
Catalog # BURK8547-1216
Supplier:   BURKLE
Threaded fittings
Catalog # BURK8547-1216
Supplier:  BURKLE
Threaded fittings
Threaded fittings

Tubing nozzle, fix, bend, external thread, nozzle-Ø 16 mm, NW 11 mm, PP, 12,7 mm

Price
38,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Gevind:
    12,7 mm
  • Beskrivelse:
    Tubing nozzle, fix, bend, external thread, nozzle-Ø 16 mm, NW 11 mm
  • Materiale:
    PP
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.